Learn More
Medchemexpress LLC Imidacloprid | 138261-41-3 | 98.45% | 200 MG
SDP
Catalog No. 5000407920
Encompass_Preferred
Imidacloprid is a potent and extensively utilized neonicotinoid pesticide employed for managing pests affecting cereals, vegetables, tea, and cotton. For research purposes, it has been shown to induce insulin resistance in cellular models.
  • Reduces phosphorylation of protein kinase B (AKT) and ribosomal S6 kinase (S6K).
  • High doses can impair cognitive functions in infant rats.
  • Developmental exposure in zebrafish leads to neurobehavioral impairments.
  • Can cause changes in antioxidant enzymes and lipid peroxidation.
  • Immunotoxic effects observed at high doses in BALB/c mice.
Catalog No. 50-004-07920 Supplier Medchemexpress LLC Supplier No. HYB0838200MG
Imidacloprid is a potent and extensively utilized neonicotinoid pesticide employed for managing pests affecting cereals, vegetables, tea, and cotton. For research purposes, it has been shown to induce insulin resistance in cellular models.
  • Reduces phosphorylation of protein kinase B (AKT) and ribosomal S6 kinase (S6K).
  • High doses can impair cognitive functions in infant rats.
  • Developmental exposure in zebrafish leads to neurobehavioral impairments.
  • Can cause changes in antioxidant enzymes and lipid peroxidation.
  • Immunotoxic effects observed at high doses in BALB/c mice.