Learn More
Cayman Chemical Imidacloprid, Purity: ≥98%, CAS: 138261-41-3, 50 mg
SDP
Catalog No. 502933176
Encompass_Preferred
A neonicotinoid insecticide; an agonist of insect nAchRs (Kis = 1.9, 2.1, 290, and 50 nM for M. persicae and D. melanogaster receptors, rat brain membranes, and rat recombinant receptors containing α4β2 subunits, respectively); lethal to M. persicae (LD95 = 2 and 160 pg/aphid via oral and topical administration, respectively); impairs learning and memory retention in honey bees at 20.8 ppb; increases body weight gain and adiposity and impairs glucose metabolism in a mouse model of high-fat diet-induced obesity at 0.06-6 mg/kg per day
Catalog No. 50-293-3176 Supplier Cayman Chemical Supplier No. 2413050 MG
A neonicotinoid insecticide; an agonist of insect nAchRs (Kis = 1.9, 2.1, 290, and 50 nM for M. persicae and D. melanogaster receptors, rat brain membranes, and rat recombinant receptors containing α4β2 subunits, respectively); lethal to M. persicae (LD95 = 2 and 160 pg/aphid via oral and topical administration, respectively); impairs learning and memory retention in honey bees at 20.8 ppb; increases body weight gain and adiposity and impairs glucose metabolism in a mouse model of high-fat diet-induced obesity at 0.06-6 mg/kg per day