Learn More
Medchemexpress LLC Imidacloprid-d4 | 1015855-75-0 | 98.9% | 1 MG
SDP
Catalog No. 5000408070
Encompass_Preferred
Imidacloprid-d4 is a deuterium-labeled form of Imidacloprid, a neonicotinoid pesticide used to control pests in various crops. This compound is intended for research use only. Stable heavy isotopes, like deuterium, are incorporated into drug molecules for use as tracers in drug development, with potential effects on pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-08070 Supplier Medchemexpress LLC Supplier No. HYB0838S1MG
Imidacloprid-d4 is a deuterium-labeled form of Imidacloprid, a neonicotinoid pesticide used to control pests in various crops. This compound is intended for research use only. Stable heavy isotopes, like deuterium, are incorporated into drug molecules for use as tracers in drug development, with potential effects on pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS