Learn More
Medchemexpress LLC Imidacloprid-d4 | 1015855-75-0 | 98.9% | 5 MG
SDP
Catalog No. 5000408160
Encompass_Preferred
Imidacloprid-d4 is the deuterium labeled version of Imidacloprid. Imidacloprid is recognized as an effective and broadly used neonicotinoid pesticide, employed for controlling pests that affect cereals, vegetables, tea, and cotton. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Application as a tracer.
  • Internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
Catalog No. 50-004-08160 Supplier Medchemexpress LLC Supplier No. HYB0838S5MG
Imidacloprid-d4 is the deuterium labeled version of Imidacloprid. Imidacloprid is recognized as an effective and broadly used neonicotinoid pesticide, employed for controlling pests that affect cereals, vegetables, tea, and cotton. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Application as a tracer.
  • Internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.