Learn More
Cayman Chemical Imidacloprid-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1015855-75-0, 10 mg
SDP
Catalog No. 502919167
Encompass_Preferred
An internal standard for the quantification of imidacloprid by GC- or LC-MS
Catalog No. 50-291-9167 Supplier Cayman Chemical Supplier No. 3954910 MG
An internal standard for the quantification of imidacloprid by GC- or LC-MS