Learn More
Cayman Chemical Imidacloprid-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1015855-75-0, 1 mg
SDP
Catalog No. 502919234
Encompass_Preferred
An internal standard for the quantification of imidacloprid by GC- or LC-MS
Catalog No. 50-291-9234 Supplier Cayman Chemical Supplier No. 395491 MG
An internal standard for the quantification of imidacloprid by GC- or LC-MS