Learn More
Cayman Chemical Imidacloprid Impurity 1, Purity: ≥98%, CAS: 105828-41-9, 500 mg
SDP
Catalog No. 502919326
Encompass_Preferred
A potential impurity in commercial preparations of imidacloprid
Catalog No. 50-291-9326 Supplier Cayman Chemical Supplier No. 37382500 MG
A potential impurity in commercial preparations of imidacloprid