Learn More
Medchemexpress LLC Imidacloprid (standard) | 138261-41-3 | 98.68% | 50 MG
SDP
Catalog No. 5000407946
Encompass_Preferred
Imidacloprid (Standard) is the analytical standard of Imidacloprid. This product is intended for research and analytical applications. Imidacloprid is an effective and widely used neonicotinoid pesticide to control pests of cereals, vegetables, tea and cotton. For research use only. We do not sell to patients.
  • Reference standard supplied assay
  • Used in qualitative, quantitative and methodological research experiments
  • Suitable for HPLC, GC and MS applications
Catalog No. 50-004-07946 Supplier Medchemexpress LLC Supplier No. HYB0838R50MG
Imidacloprid (Standard) is the analytical standard of Imidacloprid. This product is intended for research and analytical applications. Imidacloprid is an effective and widely used neonicotinoid pesticide to control pests of cereals, vegetables, tea and cotton. For research use only. We do not sell to patients.
  • Reference standard supplied assay
  • Used in qualitative, quantitative and methodological research experiments
  • Suitable for HPLC, GC and MS applications