Learn More
Medchemexpress LLC Imidacloprid (Standard) | 138261-41-3 | 98.7% | 100 MG
SDP
Catalog No. 5000407589
Encompass_Preferred
Imidacloprid (Standard) is the analytical standard of Imidacloprid, intended for research and analytical applications. It is a widely used neonicotinoid pesticide effective against pests in various crops. This product is suitable for qualitative, quantitative, and methodological research experiments.
  • Used for research and analytical applications.
  • Effective against various pests.
  • Suitable for HPLC, GC, and MS research.
  • Provided as an analytical standard.
Catalog No. 50-004-07589 Supplier Medchemexpress LLC Supplier No. HYB0838R100MG
Imidacloprid (Standard) is the analytical standard of Imidacloprid, intended for research and analytical applications. It is a widely used neonicotinoid pesticide effective against pests in various crops. This product is suitable for qualitative, quantitative, and methodological research experiments.
  • Used for research and analytical applications.
  • Effective against various pests.
  • Suitable for HPLC, GC, and MS research.
  • Provided as an analytical standard.