Learn More
Medchemexpress LLC Imidacloprid (standard) | 138261-41-3 | 98.7% | 25 MG
SDP
Catalog No. 5000450459
Encompass_Preferred
Imidacloprid (standard) is the analytical standard of Imidacloprid, intended for research and analytical applications. It is an effective and widely used neonicotinoid pesticide to control pests of cereals, vegetables, tea, and cotton.
  • Analytical standard for Imidacloprid.
  • Intended for research and analytical applications.
  • Effective and widely used neonicotinoid pesticide.
  • Used to control pests of cereals, vegetables, tea, and cotton.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-50459 Supplier Medchemexpress LLC Supplier No. 25MGHYB0838R
Imidacloprid (standard) is the analytical standard of Imidacloprid, intended for research and analytical applications. It is an effective and widely used neonicotinoid pesticide to control pests of cereals, vegetables, tea, and cotton.
  • Analytical standard for Imidacloprid.
  • Intended for research and analytical applications.
  • Effective and widely used neonicotinoid pesticide.
  • Used to control pests of cereals, vegetables, tea, and cotton.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.