Learn More
Selleck Chemical LLC Imidacloprid-urea S4448-25mg
SDP
Catalog No. 503109326
Encompass_Preferred
Imidacloprid-urea with a role as a marine xenobiotic metabolite is the primary imidacloprid soil metabolite whereas imidacloprid-olefin is the main plant-relevant metabolite and is more toxic to insects than imidacloprid
Catalog No. 50-310-9326 Supplier Selleck Chemical LLC Supplier No. S444825MG
Imidacloprid-urea with a role as a marine xenobiotic metabolite is the primary imidacloprid soil metabolite whereas imidacloprid-olefin is the main plant-relevant metabolite and is more toxic to insects than imidacloprid