Learn More
Medchemexpress LLC Imidacloprid-urea | 120868-66-8 | MFCD02661472 | 99.9% | 211.65 | C9H10ClN3O | 5 MG
SDP
Catalog No. 5000263230
Encompass_Preferred
Imidacloprid-urea is a metabolite of the neonicotinoid insecticide imidacloprid, supplied as a solid analytical reference standard for environmental fate, metabolism, and analytical method development. It is intended for laboratory research and includes reported solubility and storage recommendations to support handling and stability.
Catalog No. 50-002-63230 Supplier Medchemexpress LLC Supplier No. HY1319645MG
Imidacloprid-urea is a metabolite of the neonicotinoid insecticide imidacloprid, supplied as a solid analytical reference standard for environmental fate, metabolism, and analytical method development. It is intended for laboratory research and includes reported solubility and storage recommendations to support handling and stability.