Learn More
Medchemexpress LLC Imidacloprid-urea | 120868-66-8 | MFCD02661472 | 99.9% | C9H10ClN3O | 10MG
SDP
Catalog No. 5000261120
Encompass_Preferred
Imidacloprid-urea is the urea metabolite of the insecticide imidacloprid, provided as a high-purity analytical reference standard for research, metabolite identification, and environmental fate studies. The product is offered in small quantities and is accompanied by product documentation and batch-specific analytical data.
  • High purity suitable for analytical applications.
  • Available in multiple small sizes including 10 mg.
  • Supplied as a solid powder, off-white to light yellow.
  • Datasheet and batch-specific certificate of analysis available.
  • Intended for metabolite and environmental fate research.
Catalog No. 50-002-61120 Supplier Medchemexpress LLC Supplier No. HY13196410MG
Imidacloprid-urea is the urea metabolite of the insecticide imidacloprid, provided as a high-purity analytical reference standard for research, metabolite identification, and environmental fate studies. The product is offered in small quantities and is accompanied by product documentation and batch-specific analytical data.
  • High purity suitable for analytical applications.
  • Available in multiple small sizes including 10 mg.
  • Supplied as a solid powder, off-white to light yellow.
  • Datasheet and batch-specific certificate of analysis available.
  • Intended for metabolite and environmental fate research.