Learn More
Cayman Chemical Imidafenacin, Purity: ≥98%, CAS: 170105-16-5, 250 mg
SDP
Catalog No. 502915651
Encompass_Preferred
A muscarinic acetylcholine receptor antagonist (Kis = 7.55, 22.6, 1.42, 8.86, and 2.63 nM, respectively, for recombinant human M1-M5 receptors); binds to isolated guinea pig bladders (Kb = 0.813 nM) and inhibits ACh release from isolated rat bladders (IC50 = 0.747 nM); reduces urine volume in orally water-loaded rats from 10-300 µg/kg
Catalog No. 50-291-5651 Supplier Cayman Chemical Supplier No. 37420250 MG
A muscarinic acetylcholine receptor antagonist (Kis = 7.55, 22.6, 1.42, 8.86, and 2.63 nM, respectively, for recombinant human M1-M5 receptors); binds to isolated guinea pig bladders (Kb = 0.813 nM) and inhibits ACh release from isolated rat bladders (IC50 = 0.747 nM); reduces urine volume in orally water-loaded rats from 10-300 µg/kg