Learn More
Apexbio Technology LLC Imidapril HCl 89396-94-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000093863
Encompass_Preferred
Imidapril hydrochloride (CAS 89396-94-1) is a synthetic angiotensin-converting enzyme (ACE) inhibitor that interrupts the renin-angiotensin-aldosterone system by preventing the conversion of angiotensin I to angiotensin II This inhibition leads to vasodilation and a reduction in blood pressure and has been associated with improvements in cardiac function Imidapril HCl is extensively utilized in biomedical research to investigate the molecular mechanisms underlying hypertension and chronic heart failure as well as to evaluate ACE inhibition in various experimental models
Catalog No. 50-000-93863 Supplier Apexbio Technology LLC Supplier No. A84475.1
Imidapril hydrochloride (CAS 89396-94-1) is a synthetic angiotensin-converting enzyme (ACE) inhibitor that interrupts the renin-angiotensin-aldosterone system by preventing the conversion of angiotensin I to angiotensin II This inhibition leads to vasodilation and a reduction in blood pressure and has been associated with improvements in cardiac function Imidapril HCl is extensively utilized in biomedical research to investigate the molecular mechanisms underlying hypertension and chronic heart failure as well as to evaluate ACE inhibition in various experimental models