Learn More
Selleck Chemical LLC Imidapril HCl S2109-10mg
SDP
Catalog No. 503110015
Encompass_Preferred
Imidapril HCl(TA-6366) is an angiotensin-converting enzyme (ACE) inhibitor with IC50 of 2 6 nM used for the treatment of hypertension
Catalog No. 50-311-0015 Supplier Selleck Chemical LLC Supplier No. S210910MG
Add to Cart
Add to Cart
Imidapril HCl(TA-6366) is an angiotensin-converting enzyme (ACE) inhibitor with IC50 of 2 6 nM used for the treatment of hypertension