Learn More
Medchemexpress LLC Imidapril (hydrochloride) | 89396-94-1 | 99.9% | 1 ML
SDP
Catalog No. 5000413701
Encompass_Preferred
Imidapril hydrochloride (TA-6366) is an orally active angiotensin-converting enzyme (ACE) and MMP-9 inhibitor. It works by suppressing the conversion of angiotensin I to angiotensin II, which reduces total peripheral resistance and systemic blood pressure. This product is used for research related to hypertension, type 1 diabetic nephropathy, and chronic heart failure.
  • Orally active ACE and MMP-9 inhibitor.
  • Suppresses the conversion of angiotensin I to angiotensin II, leading to reduced total peripheral resistance and systemic blood pressure.
  • Can be used for research related to hypertension, type 1 diabetic, nephropathy, and chronic heart failure.
Catalog No. 50-004-13701 Supplier Medchemexpress LLC Supplier No. HYB145110MM/1ML
Imidapril hydrochloride (TA-6366) is an orally active angiotensin-converting enzyme (ACE) and MMP-9 inhibitor. It works by suppressing the conversion of angiotensin I to angiotensin II, which reduces total peripheral resistance and systemic blood pressure. This product is used for research related to hypertension, type 1 diabetic nephropathy, and chronic heart failure.
  • Orally active ACE and MMP-9 inhibitor.
  • Suppresses the conversion of angiotensin I to angiotensin II, leading to reduced total peripheral resistance and systemic blood pressure.
  • Can be used for research related to hypertension, type 1 diabetic, nephropathy, and chronic heart failure.