Learn More
Medchemexpress LLC Imidapril hydrochloride | 89396-94-1 | 99.9% | 25 MG
SDP
Catalog No. 5000414214
Encompass_Preferred
Imidapril hydrochloride (TA-6366) is an orally active angiotensin-converting enzyme (ACE) and MMP-9 inhibitor. It suppresses the conversion of angiotensin I to angiotensin II, leading to a reduction in total peripheral resistance and systemic blood pressure.
  • Orally active ACE and MMP-9 inhibitor
  • Reduces total peripheral resistance and systemic blood pressure
  • Used for hypertension, type 1 diabetic, nephropathy, and chronic heart failure research
Catalog No. 50-004-14214 Supplier Medchemexpress LLC Supplier No. 25MGHYB1451
Imidapril hydrochloride (TA-6366) is an orally active angiotensin-converting enzyme (ACE) and MMP-9 inhibitor. It suppresses the conversion of angiotensin I to angiotensin II, leading to a reduction in total peripheral resistance and systemic blood pressure.
  • Orally active ACE and MMP-9 inhibitor
  • Reduces total peripheral resistance and systemic blood pressure
  • Used for hypertension, type 1 diabetic, nephropathy, and chronic heart failure research