Learn More
Medchemexpress LLC Imidapril (hydrochloride) | 89396-94-1 | 99.9% | 50 MG
SDP
Catalog No. 5000413712
Encompass_Preferred
Imidapril hydrochloride (TA-6366) is an orally active angiotensin-converting enzyme (ACE) and MMP-9 inhibitor. It suppresses the conversion of angiotensin I to angiotensin II, reducing total peripheral resistance and systemic blood pressure. It is used for research related to hypertension, type 1 diabetic nephropathy, and chronic heart failure.
  • Orally active ACE inhibitor
  • MMP-9 inhibitor
  • Suppresses angiotensin I to angiotensin II conversion
  • Reduces total peripheral resistance
  • Reduces systemic blood pressure
  • Used for hypertension research
  • Used for type 1 diabetic nephropathy research
  • Used for chronic heart failure research
Catalog No. 50-004-13712 Supplier Medchemexpress LLC Supplier No. HYB145150MG
Imidapril hydrochloride (TA-6366) is an orally active angiotensin-converting enzyme (ACE) and MMP-9 inhibitor. It suppresses the conversion of angiotensin I to angiotensin II, reducing total peripheral resistance and systemic blood pressure. It is used for research related to hypertension, type 1 diabetic nephropathy, and chronic heart failure.
  • Orally active ACE inhibitor
  • MMP-9 inhibitor
  • Suppresses angiotensin I to angiotensin II conversion
  • Reduces total peripheral resistance
  • Reduces systemic blood pressure
  • Used for hypertension research
  • Used for type 1 diabetic nephropathy research
  • Used for chronic heart failure research