Learn More
Medchemexpress LLC Imidapril hydrochloride | 89396-94-1 | 99.91% | 5 MG
SDP
Catalog No. 5000413713
Encompass_Preferred
Imidapril hydrochloride (TA-6366) is an orally active angiotensin-converting enzyme (ACE) and MMP-9 inhibitor. It suppresses the conversion of angiotensin I to angiotensin II, which reduces total peripheral resistance and systemic blood pressure. It can be used for research on hypertension, type 1 diabetic nephropathy, and chronic heart failure.
  • Orally active ACE and MMP-9 inhibitor
  • Suppresses the conversion of angiotensin I to angiotensin II
  • Reduces total peripheral resistance and systemic blood pressure
  • Applicable for research in hypertension, type 1 diabetic nephropathy, and chronic heart failure
Catalog No. 50-004-13713 Supplier Medchemexpress LLC Supplier No. HYB14515MG
Imidapril hydrochloride (TA-6366) is an orally active angiotensin-converting enzyme (ACE) and MMP-9 inhibitor. It suppresses the conversion of angiotensin I to angiotensin II, which reduces total peripheral resistance and systemic blood pressure. It can be used for research on hypertension, type 1 diabetic nephropathy, and chronic heart failure.
  • Orally active ACE and MMP-9 inhibitor
  • Suppresses the conversion of angiotensin I to angiotensin II
  • Reduces total peripheral resistance and systemic blood pressure
  • Applicable for research in hypertension, type 1 diabetic nephropathy, and chronic heart failure