Learn More
Medchemexpress LLC Imidapril (hydrochloride) | 89396-94-1 | MFCD00870777 | 99.9% | 100 MG
SDP
Catalog No. 5000413700
Encompass_Preferred
Imidapril hydrochloride (TA-6366) is an orally active inhibitor of angiotensin-converting enzyme (ACE) and MMP-9. It works by suppressing the conversion of angiotensin I to angiotensin II, which in turn reduces total peripheral resistance and systemic blood pressure. It is used in research for hypertension, type 1 diabetic nephropathy, and chronic heart failure.
  • Orally active ACE and MMP-9 inhibitor
  • Suppresses conversion of angiotensin I to angiotensin II
  • Reduces total peripheral resistance and systemic blood pressure
  • Used in research for hypertension
  • Used in research for type 1 diabetic nephropathy
  • Used in research for chronic heart failure
Catalog No. 50-004-13700 Supplier Medchemexpress LLC Supplier No. HYB1451100MG
Imidapril hydrochloride (TA-6366) is an orally active inhibitor of angiotensin-converting enzyme (ACE) and MMP-9. It works by suppressing the conversion of angiotensin I to angiotensin II, which in turn reduces total peripheral resistance and systemic blood pressure. It is used in research for hypertension, type 1 diabetic nephropathy, and chronic heart failure.
  • Orally active ACE and MMP-9 inhibitor
  • Suppresses conversion of angiotensin I to angiotensin II
  • Reduces total peripheral resistance and systemic blood pressure
  • Used in research for hypertension
  • Used in research for type 1 diabetic nephropathy
  • Used in research for chronic heart failure