Learn More
Cayman Chemical Imidapril (hydrochloride), Purity: ≥98%, CAS: 89396-94-1, 10 mg
SDP
Catalog No. 502967931
Encompass_Preferred
A prodrug form of imidaprilat; inhibits swine renal cortex ACE activity in vitro (IC50 = 9.9 µM); inhibits the angiotensin I-induced pressor response in normotensive rats; decreases blood pressure in spontaneously hypertensive and 2K-1C renal hypertensive rats at 2 mg/kg; decreases stroke incidence and increases survival in stroke-prone spontaneously hypertensive rats fed a high-salt diet; reduces urinary albumin excretion in a mouse model of streptozotocin-induced diabetes at 5 mg/kg
Catalog No. 50-296-7931 Supplier Cayman Chemical Supplier No. 1885410 MG
A prodrug form of imidaprilat; inhibits swine renal cortex ACE activity in vitro (IC50 = 9.9 µM); inhibits the angiotensin I-induced pressor response in normotensive rats; decreases blood pressure in spontaneously hypertensive and 2K-1C renal hypertensive rats at 2 mg/kg; decreases stroke incidence and increases survival in stroke-prone spontaneously hypertensive rats fed a high-salt diet; reduces urinary albumin excretion in a mouse model of streptozotocin-induced diabetes at 5 mg/kg