Learn More
Medchemexpress LLC Imidazo[1,2-a]-1,3,5-triazin-4(3H)-one, 2-amino- | 67410-64-4 | 99.86% | 151.13 | 25 MG
SDP
Catalog No. 5000439488
Encompass_Preferred
5-Aza-7-deazaguanine is a substrate for wild-type (WT) E. coli purine nucleoside phosphorylase and its Ser90Ala mutant in the synthesis of base-modified nucleosides. It is for research use only.
  • For research use only
  • Substrate for wild-type (WT) E. coli purine nucleoside phosphorylase
  • Substrate for Ser90Ala mutant in the synthesis of base-modified nucleosides
Catalog No. 50-004-39488 Supplier Medchemexpress LLC Supplier No. 25MGHY111627
5-Aza-7-deazaguanine is a substrate for wild-type (WT) E. coli purine nucleoside phosphorylase and its Ser90Ala mutant in the synthesis of base-modified nucleosides. It is for research use only.
  • For research use only
  • Substrate for wild-type (WT) E. coli purine nucleoside phosphorylase
  • Substrate for Ser90Ala mutant in the synthesis of base-modified nucleosides