Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Imidazo[1,2-a]pyrazine | 274-79-3 | 99.27% | 119.13 | 1 ML
SDP

Catalog No. 5000404633
Encompass_Preferred

Imidazo[1,2-a]pyrazine is a biochemical reagent used as a biological material or organic compound for life science-related research. It is available as a solution in DMSO, with specific storage conditions to maintain its stability.

  • For research use only, not sold to patients.
  • Ships at room temperature in continental US.
  • Store powder at -20°C for 3 years or 4°C for 2 years.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month.
  • Soluble in DMSO (100 mg/mL).
  • Appearance is solid, light yellow to light brown.

Catalog No. 50-004-04633 Supplier Medchemexpress LLC Supplier No. HY3393910MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Imidazo[1,2-a]pyrazine is a biochemical reagent used as a biological material or organic compound for life science-related research. It is available as a solution in DMSO, with specific storage conditions to maintain its stability.

  • For research use only, not sold to patients.
  • Ships at room temperature in continental US.
  • Store powder at -20°C for 3 years or 4°C for 2 years.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month.
  • Soluble in DMSO (100 mg/mL).
  • Appearance is solid, light yellow to light brown.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.