Learn More
Medchemexpress LLC Imidazo[1,2-a]pyrazine, 8-(3,6-dihydro-2H-pyran-4-yl)-2-phenyl-6-[1-(4-piperidinyl)-1H-pyrazol-4-yl]- | 3023925-68-7 | C25H26N6O | 50 MG
SDP
Catalog No. 5000374133
Encompass_Preferred
YL-939 is a potent ferroptosis inhibitor that targets the PHB2/ferritin/iron axis. It efficiently protects cells from ferroptosis and has shown to ameliorate liver damage in an Acetaminophen (APAP)-induced acute liver injury model.
  • Potent ferroptosis inhibitor
  • Targets the PHB2/ferritin/iron axis
  • Protects cells from ferroptosis
  • Reduces ROS levels in cytosolic and membrane lipids
  • Inhibits ferritinophagy by improving ferritin protein expression
  • Ameliorates liver damage in APAP-induced acute liver injury models
  • High purity of 99.03%
  • Solid appearance, off-white to light yellow color
Catalog No. 50-003-74133 Supplier Medchemexpress LLC Supplier No. HY15209350MG
YL-939 is a potent ferroptosis inhibitor that targets the PHB2/ferritin/iron axis. It efficiently protects cells from ferroptosis and has shown to ameliorate liver damage in an Acetaminophen (APAP)-induced acute liver injury model.
  • Potent ferroptosis inhibitor
  • Targets the PHB2/ferritin/iron axis
  • Protects cells from ferroptosis
  • Reduces ROS levels in cytosolic and membrane lipids
  • Inhibits ferritinophagy by improving ferritin protein expression
  • Ameliorates liver damage in APAP-induced acute liver injury models
  • High purity of 99.03%
  • Solid appearance, off-white to light yellow color