Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Imidazo[1,5-a]pyridine 98.0+%, TCI America™
SDP

Catalog No. I07535G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I07535G Supplier TCI America Supplier No. I07535G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 274-47-5
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.139
MDL Number MFCD03407375
InChI Key JMANUKZDKDKBJP-UHFFFAOYSA-N
PubChem CID 274090
IUPAC Name imidazo[1,5-a]pyridine
SMILES C1=CC2=CN=CN2C=C1

Specifications

Melting Point 54°C
Quantity 5 g
Formula Weight 118.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Imidazo[1,5-a]pyridine
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.