Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Imidazo[2,1-b][1,3]thiazole-6-carboxylic acid, ≥97%, Thermo Scientific™

Catalog No. CC32501EA
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC32501EA 10 g
CC32501CB 250 mg
CC32501DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC32501EA Supplier Thermo Scientific Chemicals Supplier No. CC32501EA

Chemical Identifiers

CAS 53572-98-8
Molecular Formula C6H4N2O2S
Molecular Weight (g/mol) 168.17
MDL Number MFCD03419463
InChI Key BAKIBSRDBASFAQ-UHFFFAOYSA-N
Synonym imidazo 2,1-b thiazole-6-carboxylic acid, imidazo 2,1-b 1,3 thiazole-6-carboxylic acid, imidazo 2,1-b 1,3-thiazoline-6-carboxylic acid
PubChem CID 2759354
IUPAC Name imidazo[2,1-b][1,3]thiazole-6-carboxylic acid
SMILES C1=CSC2=NC(=CN21)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 168.18
Percent Purity 97%
Chemical Name or Material Imidazo[2, 1-b][1, 3]thiazole-6-carboxylic acid
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.