Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Imidazobenzodiazepinone derivative | 91917-65-6 | MFCD00078598 | 99.2% | 326.31 g·mol⁻¹ | C15H14N6O3 | 10 MG
SDP

Catalog No. 5000249105
Encompass_Preferred

Ro15-4513 is an imidazobenzodiazepinone derivative used in research that functions as a partial inverse agonist at GABAA (benzodiazepine) receptors and is widely used as an ethanol antagonist and reference standard.

  • Imidazobenzodiazepinone derivative acting on GABAA receptors.
  • Potent ethanol antagonist used in pharmacology and behavioral studies.
  • Off-white to yellow solid with high purity (~99.17%).
  • Molecular formula C15H14N6O3; molecular weight 326.31 g·mol⁻¹.
  • Recommended storage: powder at -20°C (long term) or 4°C (short term); in solvent at -80°C.

Catalog No. 50-002-49105 Supplier Medchemexpress LLC Supplier No. HY10347610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ro15-4513 is an imidazobenzodiazepinone derivative used in research that functions as a partial inverse agonist at GABAA (benzodiazepine) receptors and is widely used as an ethanol antagonist and reference standard.

  • Imidazobenzodiazepinone derivative acting on GABAA receptors.
  • Potent ethanol antagonist used in pharmacology and behavioral studies.
  • Off-white to yellow solid with high purity (~99.17%).
  • Molecular formula C15H14N6O3; molecular weight 326.31 g·mol⁻¹.
  • Recommended storage: powder at -20°C (long term) or 4°C (short term); in solvent at -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.