Learn More
Thermo Scientific Acetanilide Standard 2g
Supplier:  Thermo Scientific 33836700
Encompass
2 grams acetanilide powder, an organic compound C6H5NH(C=O)CH3 that is a white solid soluble in hot ethanol and ethyl acetate, used as a chemical reagent and intermediate in synthesis of pharmaceuticals and dyes.
Catalog No. 50-134-7167
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.