Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Imidazole-2-carboxylic acid

Catalog No. CDS019334 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS019334 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS019334 Supplier Sigma Aldrich Supplier No. CDS019334100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • The following aryl fluorosulfate can be utilized as a cross-coupling partner in suzuki-miyaura reactions
  • The standard reaction conditions for these palladium-catalyzed c-c bond formations are ligand-free and can be performed in water, at room temperature and are not air sensitive.palladium-catalyzed, ligand-free suzuki reaction in water using aryl fluorosulfates

Specifications

CAS 16042-25-4
Melting Point 156°C (lit.)
Molecular Formula C4H4N2O2
Linear Formula C4H4N2O2
MDL Number MFCD00270816
Quantity 500 mg
Synonym 1 H-Imidazole-2-carboxylic acid
Molecular Weight (g/mol) 112.09
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.