Learn More
Apexbio Technology LLC Imidazole 288-32-4 5g
SDP
Catalog No. 5000090259
Encompass_Preferred
Imidazole (CAS 288-32-4) is an aromatic heterocycle characterized by a five-membered ring structure containing two nitrogen atoms at the 1 and 3 positions Imidazole serves as a prototypical scaffold in medicinal chemistry notable for its role in enzyme binding and metal ion coordination due to its electron-rich nitrogen atoms It is commonly utilized in biochemical research including the study of enzyme mechanisms and as a buffer component in protein purification protocols Imidazole derivatives are also investigated as modulators of various biological targets supporting its broad relevance in pharmacological and biochemical studies
Catalog No. 50-000-90259 Supplier Apexbio Technology LLC Supplier No. B78455000
Imidazole (CAS 288-32-4) is an aromatic heterocycle characterized by a five-membered ring structure containing two nitrogen atoms at the 1 and 3 positions Imidazole serves as a prototypical scaffold in medicinal chemistry notable for its role in enzyme binding and metal ion coordination due to its electron-rich nitrogen atoms It is commonly utilized in biochemical research including the study of enzyme mechanisms and as a buffer component in protein purification protocols Imidazole derivatives are also investigated as modulators of various biological targets supporting its broad relevance in pharmacological and biochemical studies