Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Imidazole-5-propionic acid | 1074-59-5 | MFCD00237415 | 100.0% | 140.14 | C6H8N2O2 | 5 MG
SDP

Catalog No. 5000210547
Encompass_Preferred

Imidazole-5-propionic acid is a histidine metabolite supplied as a research-grade reagent for laboratory use. It is provided with analytical documentation for quality control and intended for research applications only.

  • Available in multiple sizes, including 5 mg.
  • CAS number 1074-59-5.
  • Molecular formula C6H8N2O2 and molecular weight 140.14.
  • Purity reported as 99.99% on product documentation.
  • Provided with datasheet, certificate of analysis, and safety data sheet.
  • Intended for research use only; not for human use.

Catalog No. 50-002-10547 Supplier Medchemexpress LLC Supplier No. HYW0452715MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Imidazole-5-propionic acid is a histidine metabolite supplied as a research-grade reagent for laboratory use. It is provided with analytical documentation for quality control and intended for research applications only.

  • Available in multiple sizes, including 5 mg.
  • CAS number 1074-59-5.
  • Molecular formula C6H8N2O2 and molecular weight 140.14.
  • Purity reported as 99.99% on product documentation.
  • Provided with datasheet, certificate of analysis, and safety data sheet.
  • Intended for research use only; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.