Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Imidazole, ACS, 99+%

Catalog No. AA472740E
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA472740E 2500 g
AA4727422 100 g
AA4727436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA472740E Supplier Thermo Scientific™ Supplier No. 472740E
  • $496.50
    $415.65 / Each of 1

    Save $80.85

    16% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 288-32-4
Melting Point 88-92°
Density 1.03
Boiling Point 256°
Flash Point 145°(293°F)
Molecular Formula C{3}H{4}N{2}
MDL Number MFCD00005183
Quantity 2500 g
UN Number UN3263
Merck Index 14,4912
Synonym Glyoxaline;
Formula Weight 68.08
Percent Purity 99+%
Physical Form Solid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.