Learn More
Boston Bioproducts Inc Imidazole Buffer (1 M, pH 7.0) - 500ml
Catalog No. NC1564962
Encompass
The Imidazole Buffer is an aromatic heterocyclic diazole with a buffering pH range of 6.2 to 7.8. It holds applications in molecular biology, biochemistry, and proteomics. It can be employed as a component in buffers for horseradish peroxidase assays (HRP assays), as well as a chelator for binding various divalent cations. Additionally, Imidazole aids in the elution of histidine-containing proteins from divalent cation resins, employing immobilized metal affinity chromatography (IMAC).
Catalog No. NC1564962 Supplier Boston Bioproducts Inc Supplier No. BB1117500ML
The Imidazole Buffer is an aromatic heterocyclic diazole with a buffering pH range of 6.2 to 7.8. It holds applications in molecular biology, biochemistry, and proteomics. It can be employed as a component in buffers for horseradish peroxidase assays (HRP assays), as well as a chelator for binding various divalent cations. Additionally, Imidazole aids in the elution of histidine-containing proteins from divalent cation resins, employing immobilized metal affinity chromatography (IMAC).