Learn More
Medchemexpress LLC Imidazole-d4 | 6923-01-9 | 99.9% | 100 MG
SDP
Catalog No. 5000410198
Encompass_Preferred
Imidazole-d4 is a deuterium-labeled form of Imidazole, primarily used in research. It serves as a valuable tracer and internal standard for quantitative analysis methods such as NMR, GC-MS, and LC-MS. The incorporation of stable heavy isotopes like deuterium can influence the pharmacokinetic and metabolic profiles of drug molecules, making this compound significant for drug development research.
  • Used as a tracer in various scientific applications
  • Internal standard for quantitative analysis
  • Stable heavy isotopes for drug development process
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs
  • Solid appearance, white to off-white in color
Catalog No. 50-004-10198 Supplier Medchemexpress LLC Supplier No. HYD0837S100MG
Imidazole-d4 is a deuterium-labeled form of Imidazole, primarily used in research. It serves as a valuable tracer and internal standard for quantitative analysis methods such as NMR, GC-MS, and LC-MS. The incorporation of stable heavy isotopes like deuterium can influence the pharmacokinetic and metabolic profiles of drug molecules, making this compound significant for drug development research.
  • Used as a tracer in various scientific applications
  • Internal standard for quantitative analysis
  • Stable heavy isotopes for drug development process
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs
  • Solid appearance, white to off-white in color