Learn More
Cayman Chemical Imidazole Ketone Erastin, Purity: ≥98%, CAS: 1801530-11-9, 1 mg
SDP
Catalog No. 502937074
Encompass_Preferred
An inducer of ferroptosis; inhibits glutamate release in human CCF-STTG1 astrocytoma cells (IC50 = 30 nM), indicating inhibition of the system xc- cystine/glutamate transporter; increases production of lipid ROS in SUDHL6 DLBCL cells in a concentration-dependent manner, as well as reduces GSH levels in these cells (IC50 = 34 nM); inhibits the growth of HT-1080 fibrosarcoma cells (GI50 = 310 nM) as well as HRASG12V-overexpressing BJeLR cells (IC50 = 3 nM); reduces tumor growth in an SUDHL6 mouse xenograft model at 23 and 40 mg/kg
Catalog No. 50-293-7074 Supplier Cayman Chemical Supplier No. 270881 MG
An inducer of ferroptosis; inhibits glutamate release in human CCF-STTG1 astrocytoma cells (IC50 = 30 nM), indicating inhibition of the system xc- cystine/glutamate transporter; increases production of lipid ROS in SUDHL6 DLBCL cells in a concentration-dependent manner, as well as reduces GSH levels in these cells (IC50 = 34 nM); inhibits the growth of HT-1080 fibrosarcoma cells (GI50 = 310 nM) as well as HRASG12V-overexpressing BJeLR cells (IC50 = 3 nM); reduces tumor growth in an SUDHL6 mouse xenograft model at 23 and 40 mg/kg