Learn More
Medchemexpress LLC Imidazole ketone era 10mM 1mL | 1801530-11-9 | 1 ML
SDP
Catalog No. 5000229633
Encompass_Preferred
Imidazole ketone erastin (IKE) is a selective, metabolically stable small-molecule inducer of ferroptosis that inhibits the cystine-glutamate antiporter (system xc-). It is supplied as a 10 mM solution in DMSO (1 mL) or as solid material for preparing stock solutions for cellular and in vivo research.
  • Potent inducer of ferroptosis via system xc- inhibition.
  • Provided as 10 mM solution in DMSO (1 mL) and multiple solid sizes.
  • High DMSO solubility; stock preparation volumes and concentrations provided by manufacturer.
  • Suitable for cellular assays of glutathione depletion and lipid peroxidation.
  • Store sealed away from moisture and light; in solvent store at -80°C for long-term stability.
Catalog No. 50-002-29633 Supplier Medchemexpress LLC Supplier No. HY11448110MM 1ML
Imidazole ketone erastin (IKE) is a selective, metabolically stable small-molecule inducer of ferroptosis that inhibits the cystine-glutamate antiporter (system xc-). It is supplied as a 10 mM solution in DMSO (1 mL) or as solid material for preparing stock solutions for cellular and in vivo research.
  • Potent inducer of ferroptosis via system xc- inhibition.
  • Provided as 10 mM solution in DMSO (1 mL) and multiple solid sizes.
  • High DMSO solubility; stock preparation volumes and concentrations provided by manufacturer.
  • Suitable for cellular assays of glutathione depletion and lipid peroxidation.
  • Store sealed away from moisture and light; in solvent store at -80°C for long-term stability.