Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Imidazole ketone era 5mg | 1801530-11-9 | 655.14 g/mol | C35H35ClN6O5 | 5 MG
SDP

Catalog No. 5000230032
Encompass_Preferred

Imidazole ketone erastin (IKE) is a potent, selective, and metabolically stable inhibitor of the cystine-glutamate antiporter (system xc-) that induces ferroptosis. It is supplied for research use and is used in cellular and preclinical studies of ferroptosis and tumor biology.

  • Potent inhibitor of system xc- and inducer of ferroptosis.
  • Metabolically stable analog of erastin for improved activity in biological systems.
  • High reported purity (~99.8%) suitable for research applications.
  • Molecular weight 655.14 g/mol; molecular formula C35H35ClN6O5.
  • Soluble in DMSO for preparation of concentrated stock solutions.
  • Supplied in small research quantities for laboratory experiments.

Catalog No. 50-002-30032 Supplier Medchemexpress LLC Supplier No. HY1144815MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Imidazole ketone erastin (IKE) is a potent, selective, and metabolically stable inhibitor of the cystine-glutamate antiporter (system xc-) that induces ferroptosis. It is supplied for research use and is used in cellular and preclinical studies of ferroptosis and tumor biology.

  • Potent inhibitor of system xc- and inducer of ferroptosis.
  • Metabolically stable analog of erastin for improved activity in biological systems.
  • High reported purity (~99.8%) suitable for research applications.
  • Molecular weight 655.14 g/mol; molecular formula C35H35ClN6O5.
  • Soluble in DMSO for preparation of concentrated stock solutions.
  • Supplied in small research quantities for laboratory experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.