Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Imidazole (Molecular Biology), Fisher BioReagents™,50g

Catalog No. BP30550 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
50 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP305-50 50 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP305-50
  • $652.50
    $492.65 / Each

    Save $159.85

    24% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $3,010.00
    $2,275.65 / Case of 6 EA

    Save $734.35

    24% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Commonly used buffering agent

Imidazole is an excellent biological buffer with a usable pH range of 6.2 to 7.8.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288-32-4
Molecular Formula C3H4N2
Molecular Weight (g/mol) 68.08
MDL Number MFCD00005183
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
Synonym imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole
PubChem CID 795
ChEBI CHEBI:16069
IUPAC Name 1H-imidazole
SMILES N1C=CN=C1

Specifications

Absorbance 0.01 max. (0.1M solution) at 260nm
Melting Point 86°C
Color White to Yellow
pH 10.5
Boiling Point 255°C
Assay Percent Range ≥99 %
Packaging Poly Bottle
Linear Formula HNCH:CHN:CH
Quantity 50 g
Merck Index 15, 4953
Vapor Pressure 0.003mbar at 20°C
Formula Weight 68.08
Percent Purity ≥99%
Grade DNase-, RNase- and Protease-Free
Physical Form Solid
Chemical Name or Material Imidazole
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement Signal Word: DANGER!
Precautionary Statement Emergency Overview
Causes burns by all exposure routes. Possible risk of harm to the unborn child. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artificial respiration with a respiratory device. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health: 3
Flammability: 1
Instability: 0
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.