Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Imidazole, Molecular Biology Grade, 99+%

Catalog No. AAJ6713249
Change view
Click to view available options
Quantity:
4 x 25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6713249 4 x 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6713249 Supplier Thermo Scientific™ Supplier No. J6713249
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 288-32-4 | C3H4N2 | 68.08 g/mol

Suitable for molecular biology applications

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Suitable for molecular biology applications
TRUSTED_SUSTAINABILITY

Specifications

CAS 288-32-4
Melting Point 90°
Density 1.03
Boiling Point 256°
Flash Point 145°(293°F)
Molecular Formula C3H4N2
Quantity 4 x 25 g
MDL Number MFCD00005183
UN Number UN3263
Merck Index 144,912
Synonym Glyoxaline;
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
SMILES N1C=CN=C1
IUPAC Name 1H-imidazole
Molecular Weight (g/mol) 68.08
Formula Weight 68.08
Percent Purity 99+%
Physical Form Solid
Show More Show Less
ShelfLife 5 Years
EINECSNumber 206-019-2
RTECSNumber NI3325000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.