Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Imidazole, MP Biomedicals™

Catalog No. MP21020331 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21020331 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP21020331 Supplier MP Biomedicals, Inc Supplier No. 02102033.1
Add to Cart
Add to Cart

Excellent for buffer use in pH range 6.2–7.8. Imidazole, MP Biomedicals is a chelator for the binding of various divalent cations. It is a highly polar, planar 5 membered ring that is amphoteric in nature.

Imidazole can be used for the elution of histidine containing proteins from divalent cation resins and can also be used in reverse staining of SDS-PAGE gels for detection of proteins . Imidazole is used to elute tagged proteins bound to Ni ions attached to the surface of beads in the chromatography column. Synthetic imidazoles are present in many fungicides and antifungal, antiprotozoal, and antihypertensive medications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288-32-4
Molecular Formula C3H4N2
Molecular Weight (g/mol) 68.08
MDL Number MFCD00005183
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
Synonym imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole
PubChem CID 795
ChEBI CHEBI:16069
IUPAC Name 1H-imidazole
SMILES N1C=CN=C1

Specifications

Melting Point 86°C to 94°C
Density 500–600 kg/m3 (bulk density) (Lit.)
pH ≥7 (1% aq. soln.)
Boiling Point 268°C (literature)
Quantity 100 g
Solubility Information Freely soluble in water, alcohol, ehter, chloroform, pyridine; slightly soluble in benzene; very sparingly soluble in petroleum ether.
Vapor Pressure .003hPa at 20°C (Lit.)
Formula Weight 68.1
Percent Purity ≥99%
Physical Form Crystalline Powder
Recommended Storage Store at Room Temperature (15–30°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.