Learn More
Cayman Chemical ImIdazole PropIonate 100mg
SDP
Catalog No. 502948174
Encompass_Preferred
A metabolite of histidine; decreases protein levels of IRS1, IRS2, and phosphorylated Akt and increases levels of phosphorylated p70S6K in primary mouse hepatocytes; impairs glucose tolerance in germ-free and conventionally raised mice at 500 and 100 UG/animal, respectively; plasma levels are increased in patients with type 2 diabetes
Catalog No. 50-294-8174 Supplier Cayman Chemical Supplier No. 33458100 MG
A metabolite of histidine; decreases protein levels of IRS1, IRS2, and phosphorylated Akt and increases levels of phosphorylated p70S6K in primary mouse hepatocytes; impairs glucose tolerance in germ-free and conventionally raised mice at 500 and 100 UG/animal, respectively; plasma levels are increased in patients with type 2 diabetes