Learn More
Cayman Chemical Imidazole Propionate, Purity: ≥95%, CAS: 1074-59-5, 250 mg
SDP
Catalog No. 502949028
Encompass_Preferred
A metabolite of histidine; decreases protein levels of IRS1, IRS2, and phosphorylated Akt and increases levels of phosphorylated p70S6K in primary mouse hepatocytes; impairs glucose tolerance in germ-free and conventionally raised mice at 500 and 100 µg/animal, respectively; plasma levels are increased in patients with type 2 diabetes
Catalog No. 50-294-9028 Supplier Cayman Chemical Supplier No. 33458250 MG
A metabolite of histidine; decreases protein levels of IRS1, IRS2, and phosphorylated Akt and increases levels of phosphorylated p70S6K in primary mouse hepatocytes; impairs glucose tolerance in germ-free and conventionally raised mice at 500 and 100 µg/animal, respectively; plasma levels are increased in patients with type 2 diabetes