Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Imidazole, Reagent 100 GR
SDP

Catalog No. 5026636
Encompass_Preferred

(Glyoxaline)
CAS #288-32-4 C₃H₄N₂ F.W. 68.08
Assay: min 99.5%

Imidazole is a lipid-enhancing buffer agent. When used with 4% osmium tetroxide solution in the post-fixation of tissue blocks, it produces a prominent electron-opaque staining of lipid droplets with no evidence of diffusion. Reference: Angermuller, S. Fahimi, H., Histochem. Journal 14, 823-35 (1982)

Catalog No. 50-266-36 Supplier Electron Microscopy Sciences Supplier No. 16905
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Glyoxaline)
CAS #288-32-4 C₃H₄N₂ F.W. 68.08
Assay: min 99.5%

Imidazole is a lipid-enhancing buffer agent. When used with 4% osmium tetroxide solution in the post-fixation of tissue blocks, it produces a prominent electron-opaque staining of lipid droplets with no evidence of diffusion. Reference: Angermuller, S. Fahimi, H., Histochem. Journal 14, 823-35 (1982)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.