Learn More
Gold Biotechnology Inc Imidazole Recrystallized 50 g
SDP
Catalog No. 5000562057
Encompass_Preferred
Imidazole is a planar 5-membered ring It is a highly polar highly water soluble amphoteric compound Imidazole is often used to elute tagged proteins bound to metal ions attached to the surface of beads in a chromatography column by displacing the His-tag from the metal ion and freeing the His-tagged proteins
Catalog No. 50-005-62057 Supplier Gold Biotechnology Inc Supplier No. I90250
Imidazole is a planar 5-membered ring It is a highly polar highly water soluble amphoteric compound Imidazole is often used to elute tagged proteins bound to metal ions attached to the surface of beads in a chromatography column by displacing the His-tag from the metal ion and freeing the His-tagged proteins