Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Imidazole 98.0+%, TCI America™
SDP

Catalog No. I0001500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0001500G 500 g
I000125G 25 g
I0001100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I0001500G Supplier TCI America Supplier No. I0001500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 288-32-4
Molecular Formula C3H4N2
Molecular Weight (g/mol) 68.08
MDL Number MFCD00005183
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
Synonym imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole
PubChem CID 795
ChEBI CHEBI:16069
IUPAC Name 1H-imidazole
SMILES N1C=CN=C1

Specifications

Melting Point 90°C
Color White
Boiling Point 256°C
Quantity 500 g
UN Number 2923
Formula Weight 68.08
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Imidazole
EINECSNumber (5)-0381
RTECSNumber NI3325000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.