Learn More
Medchemexpress LLC Imidazoleacetic acid | 645-65-8 | 99.8% | 126.11 | 10 MG
SDP
Catalog No. 5000436575
Encompass_Preferred
Imidazoleacetic acid is a blood-brain barrier-permeable full agonist of the GABAA receptor. It is formed via histamine oxidation in the mouse brain and exerts multiple neurochemical and behavioral effects.
  • Acts as a full agonist of the GABAA receptor
  • Formed via histamine oxidation in the mouse brain
  • Exerts multiple neurochemical effects
  • Induces analgesia, sedation, and hypnosis
  • Reduces blood pressure and body temperature
  • Decreases isolation-induced aggression and motor activity
Catalog No. 50-004-36575 Supplier Medchemexpress LLC Supplier No. HY11341310MG
Imidazoleacetic acid is a blood-brain barrier-permeable full agonist of the GABAA receptor. It is formed via histamine oxidation in the mouse brain and exerts multiple neurochemical and behavioral effects.
  • Acts as a full agonist of the GABAA receptor
  • Formed via histamine oxidation in the mouse brain
  • Exerts multiple neurochemical effects
  • Induces analgesia, sedation, and hypnosis
  • Reduces blood pressure and body temperature
  • Decreases isolation-induced aggression and motor activity