Learn More
Medchemexpress LLC Imidazoleacetic acid hydrochloride | 3251-69-2 | 162.58 | 10 G
SDP
Catalog No. 5000446552
Encompass_Preferred
Imidazoleacetic acid hydrochloride is a γ-aminobutyric acid receptor agonist that non-competitively inhibits GABA-T, increasing total free GABA in the brain. It is intended for research use only.
  • Acts as a γ-aminobutyric acid receptor agonist
  • Non-competitively inhibits GABA-T with a Ki value of 0.34 mM
  • Increases total free GABA in the brain
  • Suitable for in vitro and in vivo research applications
Catalog No. 50-004-46552 Supplier Medchemexpress LLC Supplier No. HYW00768610G
Imidazoleacetic acid hydrochloride is a γ-aminobutyric acid receptor agonist that non-competitively inhibits GABA-T, increasing total free GABA in the brain. It is intended for research use only.
  • Acts as a γ-aminobutyric acid receptor agonist
  • Non-competitively inhibits GABA-T with a Ki value of 0.34 mM
  • Increases total free GABA in the brain
  • Suitable for in vitro and in vivo research applications