Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Imidazolidinyl urea

Catalog No. I5133100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I5133100G 100 g
I513325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I5133100G Supplier Sigma Aldrich Supplier No. I5133100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used in:Pharmacological studies investigating nutrient-sensitized screening for drugs that shift energy metabolism from mitochondrial respiration to glycolysisTherapeutic studies involving nanostructured vegetable-based carriers for topical delivery of active moleculesTopical formulas for use as antioxidants

Specifications

CAS 39236-46-9
Molecular Formula C11H16N8O8
Linear Formula C11H16N8O8
MDL Number MFCD00221482
Quantity 100 g
Synonym N,N-Methylenebis[N-[3-(hydroxymethyl)-2,5-dioxo-4-imidazolidinyl]-urea
Molecular Weight (g/mol) 388.29
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.