Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Imidodicarbonimidic diamide, N-methyl-, hydrochloride (1:1) | 1674-62-0 | ≥97.0% | 151.60 | 100 MG
SDP

Catalog No. 5000400929
Encompass_Preferred

Metformin impurity 1 is a chemical product from MedChemExpress, identified by CAS number 1674-62-0 and molecular weight 151.60. It is intended for research use in laboratory settings.

  • White to off-white solid appearance.
  • Purity of at least 97.0%.
  • Stable under recommended storage conditions.
  • Melting point range of 148-151 °C.
  • Recommended storage at 4°C, sealed, away from moisture and light.
  • Harmful if swallowed.
  • For research use only.

Catalog No. 50-004-00929 Supplier Medchemexpress LLC Supplier No. HY23875100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Metformin impurity 1 is a chemical product from MedChemExpress, identified by CAS number 1674-62-0 and molecular weight 151.60. It is intended for research use in laboratory settings.

  • White to off-white solid appearance.
  • Purity of at least 97.0%.
  • Stable under recommended storage conditions.
  • Melting point range of 148-151 °C.
  • Recommended storage at 4°C, sealed, away from moisture and light.
  • Harmful if swallowed.
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.